Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRUNE2 SIRNA Assays ›
Gene Symbol:
PRUNE2
Gene Name:
prune homolog 2
Gene Aliases:
BMCC1, BNIPXL, C9orf65, KIAA0367
Chromosome Location:
Chr.9: 76611376 - 76906220 on Build GRCh38
UniGene ID:
Hs.262857
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
36985
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRUNE2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308047.1
NP_001294976.1
3
350
NM_001308048.1
NP_001294977.1
3
350
NM_015225.2
NP_056040.2
3
350
XM_005251745.1
XP_005251802.1
3
483
XM_005251746.1
XP_005251803.1
3
483
XM_005251748.1
XP_005251805.1
3
483
XM_005251750.1
XP_005251807.1
3
483
XM_005251751.1
XP_005251808.1
3
483
XM_005251754.1
XP_005251811.1
3
483
XM_006716982.1
XP_006717045.1
3
483
XM_006716983.1
XP_006717046.1
3
483
XM_006716984.2
XP_006717047.1
3
483
XM_006716985.1
XP_006717048.1
3
483
XM_006716986.1
XP_006717049.1
3
483
XM_011518323.1
XP_011516625.1
3
483
XM_011518326.2
XP_011516628.1
3
483
XM_011518327.1
XP_011516629.1
3
483
XM_011518328.2
XP_011516630.1
3
483
XM_017014345.1
XP_016869834.1
3
483
XM_017014346.1
XP_016869835.1
3
483
XM_017014347.1
XP_016869836.1
3
483
XM_017014348.1
XP_016869837.1
3
483
XM_017014349.1
XP_016869838.1
3
483
XM_017014350.1
XP_016869839.1
3
483
XM_017014351.1
XP_016869840.1
3
483
XM_017014352.1
XP_016869841.1
3
483
XM_017014353.1
XP_016869842.1
3
483
GenBank mRNA
AB535152.1
-
3
226
BC019095.2
-
3
350
FJ808772.1
-
3
304
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
pyrophosphatase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline