Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: A2ML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282424.2 | NP_001269353.1 | 22 | 2882 | ||
NM_144670.5 | NP_653271.2 | 33 | 4367 | |||
XM_011520566.2 | XP_011518868.1 | 33 | 4324 | |||
XM_011520567.2 | XP_011518869.1 | 33 | 4324 | |||
XM_017018868.1 | XP_016874357.1 | 33 | 4321 | |||
XM_017018869.1 | XP_016874358.1 | 33 | 4321 | |||
XM_017018870.1 | XP_016874359.1 | 33 | 4285 | |||
GenBank mRNA | AK057908.1 | - | 4 | 369 | ||
AK096448.1 | - | 11 | 1356 | |||
AK122624.1 | - | 27 | 3453 | |||
AK123591.1 | - | 16 | 2020 | |||
AK123592.1 | - | 22 | 2837 | |||
AK123601.1 | - | 30 | 3744 | |||
AK290355.1 | - | 33 | 4220 | |||
AK302555.1 | - | 22 | 2882 | |||
AL832139.1 | - | 33 | 4218 | |||
AL832750.1 | - | 11 | 1092 | |||
AL833479.1 | - | 31 | 3883 | |||
BC093840.1 | - | 4 | 320 | |||
BC112131.1 | - | 4 | 320 | |||
BX647674.1 | - | 15 | 1822 | |||
BX648231.1 | - | 34 | 4219 |