Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BTBD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017523.1 | NP_001017523.1 | 15 | 2437 | ||
NM_001018072.1 | NP_001018082.1 | 17 | 4297 | |||
XM_005268645.3 | XP_005268702.1 | 17 | 2665 | |||
XM_011537909.2 | XP_011536211.1 | 17 | 3220 | |||
XM_011537910.2 | XP_011536212.1 | 17 | 3174 | |||
XM_017018807.1 | XP_016874296.1 | 16 | 5447 | |||
GenBank mRNA | AK091276.1 | - | 7 | 1272 | ||
AK094304.1 | - | 1 | 429 | |||
AK122818.1 | - | 17 | 2734 | |||
AK127012.1 | - | 14 | 2207 | |||
AK127295.1 | - | 15 | 2437 | |||
AK128445.1 | - | 9 | 1676 | |||
AY373588.1 | - | 13 | 2131 |