Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC44A3 SIRNA Assays ›
Gene Symbol:
SLC44A3
Gene Name:
solute carrier family 44 member 3
Gene Aliases:
CTL3
Chromosome Location:
Chr.1: 94820342 - 94895247 on Build GRCh38
UniGene ID:
Hs.483423
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
38616
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC44A3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114106.2
NP_001107578.1
3
379
NM_001258340.1
NP_001245269.1
3
379
NM_001258341.1
NP_001245270.1
3
379
NM_001258342.1
NP_001245271.1
2
271
NM_001258343.1
NP_001245272.1
2
271
NM_001301079.1
NP_001288008.1
2
314
NM_152369.4
NP_689582.2
2
314
XM_005270440.2
XP_005270497.1
3
370
XM_005270441.2
XP_005270498.1
3
316
XM_005270442.1
XP_005270499.1
3
370
XM_011540632.2
XP_011538934.1
1
133
XM_011540633.2
XP_011538935.1
1
67
XM_011540635.2
XP_011538937.1
3
370
XM_011540636.2
XP_011538938.1
3
370
XM_017000262.1
XP_016855751.1
2
256
XM_017000263.1
XP_016855752.1
3
316
XM_017000264.1
XP_016855753.1
3
370
XM_017000265.1
XP_016855754.1
3
370
XM_017000266.1
XP_016855755.1
3
370
GenBank mRNA
AK300193.1
-
2
208
AK301288.1
-
2
216
AK303906.1
-
2
250
AK313574.1
-
3
267
AK316123.1
-
4
320
AL530322.3
-
3
371
AL540829.3
-
2
271
BC033858.1
-
3
342
BC053877.1
-
2
237
5760394
BX648225.1
-
1
133
DA538278.1
-
2
314
DC311042.1
-
3
376
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylcholine biosynthetic process
choline transport
transmembrane transport
protein binding
choline transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-pkth9:80/10.239.167.245:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0