Search Thermo Fisher Scientific
Gene Symbol: C5orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135586.1 | NP_001129058.1 | 2 | 683 | ||
NM_152409.3 | NP_689622.2 | 2 | 496 | |||
XM_005271889.3 | XP_005271946.1 | 2 | 1332 | |||
XM_017009049.1 | XP_016864538.1 | 2 | 851 | |||
XM_017009050.1 | XP_016864539.1 | 2 | 459 | |||
GenBank mRNA | AK094881.1 | - | 2 | 633 | ||
BC053677.1 | - | 2 | 496 | 6166677 | ||
BX648529.1 | - | 2 | 563 |