Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LCA5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152505.3 | NP_689718.1 | 4 | 421 | ||
XM_005260926.1 | XP_005260983.1 | 4 | 379 | |||
XM_006723967.1 | XP_006724030.1 | 3 | 424 | |||
XM_011529458.1 | XP_011527760.1 | 5 | 830 | |||
XM_011529459.1 | XP_011527761.1 | 5 | 489 | |||
XM_011529460.1 | XP_011527762.1 | 6 | 629 | |||
XM_011529461.1 | XP_011527763.1 | 6 | 736 | |||
XM_011529462.1 | XP_011527764.1 | 4 | 379 | |||
XM_011529463.2 | XP_011527765.1 | 4 | 432 | |||
XM_011529464.1 | XP_011527766.1 | 5 | 458 | |||
XM_011529465.1 | XP_011527767.1 | 4 | 377 | |||
XM_011529466.1 | XP_011527768.1 | 4 | 615 | |||
XM_011529467.1 | XP_011527769.1 | 5 | 693 | |||
XM_011529468.1 | XP_011527770.1 | 4 | 335 | |||
XM_011529469.2 | XP_011527771.1 | 4 | 722 | |||
XM_011529470.1 | XP_011527772.1 | 3 | 218 | |||
XM_011529471.1 | XP_011527773.1 | 3 | 424 | |||
GenBank mRNA | BC031059.2 | - | 4 | 422 | 5271223 | |
BC043006.1 | - | 4 | 638 | 5297703 | ||
BX647793.1 | - | 3 | 456 | |||
BX648744.1 | - | 1 | 49 |