Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other THRB SIRNA Assays ›
Gene Symbol:
THRB
Gene Name:
thyroid hormone receptor beta
Gene Aliases:
C-ERBA-2, C-ERBA-BETA, ERBA2, GRTH, NR1A2, PRTH, THR1, THRB1, THRB2
Chromosome Location:
Chr.3: 24117153 - 24495281 on Build GRCh38
UniGene ID:
Hs.187861
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
3918
Purification
In-Vivo Ready
Standard
HPLC
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
417.00
Your Price
Online offer:
417.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: THRB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000461.4
NP_000452.2
4
427
NM_001128176.2
NP_001121648.1
5
538
NM_001128177.1
NP_001121649.1
5
564
NM_001252634.1
NP_001239563.1
6
662
XM_005265421.4
XP_005265478.1
5
635
XM_005265423.4
XP_005265480.1
4
470
XM_005265424.3
XP_005265481.1
4
405
XM_006713317.3
XP_006713380.1
5
778
XM_006713318.3
XP_006713381.1
5
730
XM_011534046.2
XP_011532348.1
5
2678
XM_011534047.2
XP_011532349.1
6
915
XM_011534048.2
XP_011532350.1
5
494
XM_011534049.2
XP_011532351.1
4
350
XM_011534050.2
XP_011532352.1
5
723
XM_011534051.2
XP_011532353.1
4
313
XM_011534052.2
XP_011532354.1
5
450
XM_011534053.2
XP_011532355.1
4
571
XM_017007108.1
XP_016862597.1
6
848
XM_017007109.1
XP_016862598.1
7
1006
XM_017007110.1
XP_016862599.1
6
813
XM_017007111.1
XP_016862600.1
5
459
XM_017007112.1
XP_016862601.1
4
429
XM_017007113.1
XP_016862602.1
3
568
XM_017007114.1
XP_016862603.1
3
533
XM_017007115.1
XP_016862604.1
3
407
GenBank mRNA
AB307687.1
-
1
127
AK096628.1
-
6
651
AY286465.1
-
4
353
AY286466.1
-
4
371
AY286467.1
-
4
337
AY286468.1
-
5
456
AY286469.1
-
5
447
AY286470.1
-
5
481
AY286471.1
-
6
530
BC106929.1
-
3
252
40033198
BC106930.1
-
4
398
40033200
BP313943.1
-
4
416
BQ879477.1
-
4
549
DA218319.1
-
4
367
GQ919288.1
-
4
290
HQ692825.1
-
2
142
M26747.1
-
4
427
X04707.1
-
4
427
Back To Top
More Information
Panther Classification:
Molecular Function -
C4 zinc finger nuclear receptor
DNA-binding transcription factor
zinc finger transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
sensory perception of sound
negative regulation of female receptivity
regulation of heart contraction
female courtship behavior
organ morphogenesis
intracellular receptor signaling pathway
steroid hormone mediated signaling pathway
positive regulation of transcription from RNA polymerase II promoter
Type I pneumocyte differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
transcription corepressor activity
thyroid hormone receptor activity
protein binding
zinc ion binding
enzyme binding
chromatin DNA binding
sequence-specific DNA binding
thyroid hormone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-zl7kt:80/100.66.73.1:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0