Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other USP54 SIRNA Assays ›
          Gene Symbol:
          USP54
          Gene Name:
          ubiquitin specific peptidase 54
          Gene Aliases:
          C10orf29, bA137L10.3, bA137L10.4
          Chromosome Location:
          Chr.10: 73497538 - 73626844 on Build GRCh38
          UniGene ID:
          Hs.657355
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 39360

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: USP54
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320437.1 NP_001307366.1 18 3380
          NM_152586.3 NP_689799.3 18 3478
          NR_135249.1 19 3865
          NR_135250.1 14 2860
          XM_005269582.2 XP_005269639.1 17 6556
          XM_011539368.2 XP_011537670.1 18 6622
          XM_017015759.1 XP_016871248.1 20 6051
          XM_017015760.1 XP_016871249.1 20 4953
          XM_017015761.1 XP_016871250.1 20 5791
          XM_017015762.1 XP_016871251.1 20 4961
          XM_017015763.1 XP_016871252.1 20 4255
          XM_017015764.1 XP_016871253.1 20 4318
          XM_017015765.1 XP_016871254.1 21 4571
          XM_017015766.1 XP_016871255.1 20 6177
          XM_017015767.1 XP_016871256.1 20 5836
          XM_017015768.1 XP_016871257.1 20 4007
          XM_017015769.1 XP_016871258.1 20 3615
          XM_017015770.1 XP_016871259.1 20 4276
          XM_017015771.1 XP_016871260.1 19 6793
          XM_017015772.1 XP_016871261.1 19 6799
          XM_017015773.1 XP_016871262.1 18 6733
          XM_017015774.1 XP_016871263.1 19 5719
          XM_017015775.1 XP_016871264.1 20 5597
          XM_017015776.1 XP_016871265.1 19 6102
          XM_017015777.1 XP_016871266.1 19 5761
          XM_017015778.1 XP_016871267.1 18 6679
          XM_017015779.1 XP_016871268.1 19 6799
          XM_017015780.1 XP_016871269.1 19 6799
          XM_017015781.1 XP_016871270.1 18 6628
          XM_017015782.1 XP_016871271.1 19 4379
          XM_017015783.1 XP_016871272.1 19 5426
          XM_017015784.1 XP_016871273.1 18 3372
          XM_017015785.1 XP_016871274.1 18 6679
          XM_017015786.1 XP_016871275.1 18 3869
          GenBank mRNA AK094637.1 - 1 572
          AK126497.1 - 6 1833
          AK127213.1 - 5 1413
          AK127267.1 - 14 2860
          AK303633.1 - 3 1071
          AL832102.1 - 1 518
          BC026037.1 - 1 326
          BC064967.1 - 1 514
          BC130633.1 - 3 1062

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          protein deubiquitination
          protein binding
          thiol-dependent ubiquitinyl hydrolase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0