Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C15orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301106.1 | NP_001288035.1 | 2 | 947 | ||
NM_153040.2 | NP_694585.1 | 2 | 947 | |||
XM_011521257.2 | XP_011519559.1 | 2 | 6862 | |||
GenBank mRNA | AK057393.1 | - | 2 | 948 | ||
BC109252.2 | - | 2 | 703 | 40023840 | ||
BC109253.1 | - | 2 | 701 | 40023844 |