Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TANGO2 SIRNA Assays ›
Gene Symbol:
TANGO2
Gene Name:
transport and golgi organization 2 homolog
Gene Aliases:
C22orf25, MECRCN
Chromosome Location:
Chr.22: 20017000 - 20067164 on Build GRCh38
UniGene ID:
Hs.474233
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
39921
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TANGO2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283106.2
NP_001270035.1
10
1721
NM_001283116.2
NP_001270045.1
9
1475
NM_001283129.2
NP_001270058.1
9
1715
NM_001283148.2
NP_001270077.1
9
1473
NM_001283154.2
NP_001270083.1
9
1585
NM_001283179.2
NP_001270108.1
7
1416
NM_001283186.2
NP_001270115.1
7
1289
NM_001283199.2
NP_001270128.1
7
1362
NM_001283215.2
NP_001270144.1
7
1456
NM_001283235.2
NP_001270164.1
8
1360
NM_001283248.2
NP_001270177.1
6
1135
NM_001322141.1
NP_001309070.1
9
1871
NM_001322142.1
NP_001309071.1
9
1631
NM_001322143.1
NP_001309072.1
9
1788
NM_001322144.1
NP_001309073.1
9
1713
NM_001322145.1
NP_001309074.1
7
1602
NM_001322146.1
NP_001309075.1
9
1781
NM_001322147.1
NP_001309076.1
7
1529
NM_001322148.1
NP_001309077.1
9
1792
NM_001322149.1
NP_001309078.1
7
1490
NM_001322150.1
NP_001309079.1
8
1756
NM_001322153.1
NP_001309082.1
8
1516
NM_001322155.1
NP_001309084.1
8
1589
NM_001322160.1
NP_001309089.1
9
1675
NM_001322163.1
NP_001309092.1
8
1535
NM_001322166.1
NP_001309095.1
7
1362
NM_001322167.1
NP_001309096.1
7
1414
NM_001322169.1
NP_001309098.1
7
1287
NM_001322171.1
NP_001309100.1
8
1433
NM_001322172.1
NP_001309101.1
8
1725
NM_001322173.1
NP_001309102.1
8
1600
NM_001322174.1
NP_001309103.1
8
1487
NM_001322175.1
NP_001309104.1
8
1485
NM_152906.6
NP_690870.3
9
1587
NR_104274.2
7
1206
NR_104275.2
7
1446
NR_136206.1
7
1385
NR_136211.1
9
1571
NR_136212.1
8
1467
XM_011529863.1
XP_011528165.1
9
1624
XM_011529865.1
XP_011528167.1
7
1337
XM_011529867.1
XP_011528169.1
9
1570
XM_017028577.1
XP_016884066.1
9
1704
XM_017028578.1
XP_016884067.1
9
1723
XM_017028579.1
XP_016884068.1
9
1752
XM_017028580.1
XP_016884069.1
10
1841
XM_017028581.1
XP_016884070.1
9
1835
XM_017028582.1
XP_016884071.1
10
1918
XM_017028583.1
XP_016884072.1
9
1665
XM_017028584.1
XP_016884073.1
9
1928
XM_017028585.1
XP_016884074.1
7
1517
XM_017028586.1
XP_016884075.1
7
1442
XM_017028587.1
XP_016884076.1
9
1679
XM_017028588.1
XP_016884077.1
8
1374
GenBank mRNA
AK092484.1
-
7
1244
AL713640.1
-
6
1215
BC041339.1
-
9
1550
5284126
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-jgtjs:80/10.239.162.121:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0