Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: PSTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153336.2 | NP_699167.2 | 4, 5 | 1231 | ||
XM_017015640.1 | XP_016871129.1 | 4, 5 | 857 | |||
GenBank mRNA | AK094271.1 | - | 3, 4 | 1336 | ||
BC035344.1 | - | 4, 5 | 1231 | 5185692 |