Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CACUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153810.4 | NP_722517.3 | 9 | 2016 | ||
GenBank mRNA | AY743663.1 | - | 9 | 2017 | ||
BC038294.2 | - | 10 | 2064 | 4837841 |