Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002617.3 | NP_002608.1 | 6 | 1311 | ||
NM_153818.1 | NP_722540.1 | 6 | 1371 | |||
XM_011541573.1 | XP_011539875.1 | 6 | 1419 | |||
GenBank mRNA | AB013818.1 | - | 6 | 1311 | ||
AK124816.1 | - | 6 | 1289 | |||
BC000543.2 | - | 6 | 1330 | 3163782 | ||
BC018198.1 | - | 6 | 1293 | 3882695 |