Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RARB SIRNA Assays ›
          Gene Symbol:
          RARB
          Gene Name:
          retinoic acid receptor beta
          Gene Aliases:
          HAP, MCOPS12, NR1B2, RARbeta1, RRB2
          Chromosome Location:
          Chr.3: 24829323 - 25597932 on Build GRCh38
          UniGene ID:
          Hs.543218
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 4124

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RARB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000965.4 NP_000956.2 8 1945
          NM_001290216.2 NP_001277145.1 11 1988
          NM_001290217.1 NP_001277146.1 8 1403
          NM_001290266.1 NP_001277195.1 8 1588
          NM_001290276.1 NP_001277205.1 8 1945
          NM_001290277.1 NP_001277206.1 8 1807
          NM_001290300.1 NP_001277229.1 8 1409
          NM_016152.3 NP_057236.1 8 1588
          NR_110892.1 7 1784
          NR_110893.1 7 1740
          GenBank mRNA AK299756.1 - 9 1521
          AY501390.1 - 8 1377
          BC050415.2 - 6 1171
          BC060794.1 - 8 1627 30341884
          Y00291.1 - 8 1797

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031529.
          This siRNA also targets the following transcripts in Mouse: NM_001289761, NM_011243, NM_001289760, NM_001289762, S56660, and others.

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ureteric bud development
          glandular epithelial cell development
          outflow tract septum morphogenesis
          retinal pigment epithelium development
          growth plate cartilage development
          transcription initiation from RNA polymerase II promoter
          signal transduction
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          striatum development
          regulation of myelination
          negative regulation of chondrocyte differentiation
          embryonic hindlimb morphogenesis
          multicellular organism growth
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          steroid hormone mediated signaling pathway
          positive regulation of neuron differentiation
          positive regulation of transcription from RNA polymerase II promoter
          embryonic eye morphogenesis
          retinoic acid receptor signaling pathway
          embryonic digestive tract development
          ventricular cardiac muscle cell differentiation
          RNA polymerase II regulatory region sequence-specific DNA binding
          DNA binding
          steroid hormone receptor activity
          retinoic acid receptor activity
          drug binding
          zinc ion binding
          protein complex binding
          retinoid X receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0