siRNA Details
Gene Symbol: GBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301687.1 | NP_001288616.1 | 3 | 627 | ||
NM_001485.3 | NP_001476.2 | 2 | 590 | |||
GenBank mRNA | AF118452.1 | - | 2 | 590 | ||
AK311262.1 | - | 2 | 221 | |||
BC137448.1 | - | 2 | 590 | 9021448 | ||
BC137449.1 | - | 2 | 590 | 9021449 | ||
BC144646.1 | - | 3 | 627 |

More Information
Other Species Targeted

Panther Classification:
