Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GYPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304382.1 | NP_001291311.1 | 3, 4 | 275 | ||
NM_002100.5 | NP_002091.3 | 2, 3 | 195 | |||
XM_011531903.2 | XP_011530205.1 | 2, 3 | 330 | |||
XM_011531904.2 | XP_011530206.1 | 3, 4 | 2834 | |||
XM_011531905.2 | XP_011530207.1 | 3, 4 | 482 | |||
XM_017008137.1 | XP_016863626.1 | 2, 3 | 2529 | |||
GenBank mRNA | AY950609.1 | - | 2, 3 | 137 | ||
AY950611.1 | - | 2, 3 | 137 | |||
BC069310.1 | - | 2, 3 | 187 | 7262104 | ||
BC121077.1 | - | 2, 3 | 171 | 40119713 | ||
BC121078.1 | - | 2, 3 | 171 | 40119716 | ||
EU338224.1 | - | 3, 4 | 298 | |||
EU338226.1 | - | 3, 4 | 232 | |||
EU338229.1 | - | 3, 4 | 298 | |||
EU338230.1 | - | 3, 4 | 298 | |||
EU338236.1 | - | 3, 4 | 298 | |||
J02982.1 | - | 2, 3 | 147 | |||
X08055.1 | - | 4, 5 | 273 |