Search
Search

NDUFC1| Gene Symbol: NDUFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184986.1 | NP_001171915.1 | 6 | 1189 | ||
| NM_001184987.1 | NP_001171916.1 | 5 | 657 | |||
| NM_001184988.1 | NP_001171917.1 | 6 | 693 | |||
| NM_001184989.1 | NP_001171918.1 | 6 | 716 | |||
| NM_001184990.1 | NP_001171919.1 | 5 | 653 | |||
| NM_001184991.1 | NP_001171920.1 | 5 | 1130 | |||
| NM_002494.3 | NP_002485.1 | 5 | 792 | |||
| GenBank mRNA | AF047184.1 | - | 4 | 387 | ||
| AF047435.1 | - | 4 | 367 | |||
| AK291147.1 | - | 4 | 397 | |||
| BC107682.1 | - | 5 | 762 | 5557684 | ||
| CN347371.1 | - | 5 | 558 | |||