Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UCN SIRNA Assays ›
Gene Symbol:
UCN
Gene Name:
urocortin
Gene Aliases:
UI, UROC
Chromosome Location:
Chr.2: 27307400 - 27308445 on Build GRCh38
UniGene ID:
Hs.534363
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
41856
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UCN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003353.3
NP_003344.1
2
646
GenBank mRNA
BC104470.1
-
2
447
40033510
BC104471.2
-
2
447
40033515
CR542244.1
-
1
347
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_019150, U33935.
This siRNA also targets the following transcripts in Mouse: NM_021290, BC144891, BC116948, BC116950.
Panther Classification:
Molecular Function -
peptide hormone
Gene Ontology Categories:
Function(s)
Process(es)
startle response
inflammatory response
response to oxidative stress
G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
female pregnancy
sensory perception of sound
associative learning
aerobic respiration
negative regulation of gene expression
response to auditory stimulus
pancreatic juice secretion
positive regulation of cell growth
positive regulation of cAMP biosynthetic process
negative regulation of histone deacetylation
neuron projection development
negative regulation of appetite
response to estradiol
positive regulation of interleukin-6 production
positive regulation of collagen biosynthetic process
positive regulation of peptidyl-serine phosphorylation
activation of protein kinase A activity
social behavior
gastric emptying
drinking behavior
negative regulation of apoptotic process
positive regulation of vascular permeability
positive regulation of translation
positive regulation of DNA replication
negative regulation of blood pressure
negative regulation of cell size
positive regulation of vasodilation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of hormone secretion
response to pain
response to glucocorticoid
positive regulation of corticotropin secretion
regulation of synaptic transmission, glutamatergic
positive regulation of cardiac muscle contraction
negative regulation of gastric acid secretion
negative regulation of necrotic cell death
positive regulation of calcium ion import
negative regulation of neuron death
negative regulation of feeding behavior
positive regulation of behavioral fear response
neuropeptide hormone activity
protein binding
corticotropin-releasing hormone activity
histone deacetylase inhibitor activity
corticotropin-releasing hormone receptor 1 binding
corticotropin-releasing hormone receptor 2 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0