Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AURKA SIRNA Assays ›
          Gene Symbol:
          AURKA
          Gene Name:
          aurora kinase A
          Gene Aliases:
          AIK, ARK1, AURA, BTAK, PPP1R47, STK15, STK6, STK7
          Chromosome Location:
          Chr.20: 56369389 - 56392337 on Build GRCh38
          UniGene ID:
          Hs.250822
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 425

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 6,301.00
          Your Price
          Online offer:
          6301.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AURKA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001323303.1 NP_001310232.1 4 579
          NM_001323304.1 NP_001310233.1 4 342
          NM_001323305.1 NP_001310234.1 5 452
          NM_003600.3 NP_003591.2 3 469
          NM_198433.2 NP_940835.1 5 677
          NM_198434.2 NP_940836.1 4 368
          NM_198435.2 NP_940837.1 3 258
          NM_198436.2 NP_940838.1 4 354
          NM_198437.2 NP_940839.1 3 244
          XM_017028034.1 XP_016883523.1 5 799
          XM_017028035.1 XP_016883524.1 3 159
          GenBank mRNA AF008551.1 - 2 69
          AF011468.1 - 3 325
          AK301769.1 - 3 135
          AK312776.1 - 3 136
          AL711075.1 - 3 337
          BC001280.1 - 4 201 3456017
          BC002499.2 - 3 197 3051177
          BC006423.1 - 3 94 3532438
          BC027464.1 - 4 236 4449474
          BF029182.1 - 125
          BM458030.1 - 4 269
          BQ435059.1 - 3 90
          BU171303.1 - 3 190
          BU542363.1 - 5 529
          CB113289.1 - 5 371
          CB138172.1 - 4 311
          D84212.1 - 4 238

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          meiotic spindle organization
          protein phosphorylation
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          spindle organization
          mitotic spindle organization
          spindle assembly involved in female meiosis I
          mitotic nuclear division
          mitotic centrosome separation
          anterior/posterior axis specification
          anaphase-promoting complex-dependent catabolic process
          regulation of protein stability
          negative regulation of protein binding
          response to estradiol
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          regulation of cytokinesis
          histone-serine phosphorylation
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic process
          positive regulation of mitotic nuclear division
          regulation of centrosome cycle
          protein autophosphorylation
          cell division
          centrosome localization
          protein localization to centrosome
          positive regulation of oocyte maturation
          regulation of signal transduction by p53 class mediator
          neuron projection extension
          protein kinase activity
          protein serine/threonine kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein binding
          ATP binding
          protein kinase binding
          ubiquitin protein ligase binding
          histone serine kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-97z6t:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0