Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POU6F1 SIRNA Assays ›
Gene Symbol:
POU6F1
Gene Name:
POU class 6 homeobox 1
Gene Aliases:
BRN5, MPOU, TCFB1
Chromosome Location:
Chr.12: 51186936 - 51218134 on Build GRCh38
UniGene ID:
Hs.555886
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4251
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POU6F1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002702.3
NP_002693.3
5
907
NR_026893.1
10
1365
XM_017019507.1
XP_016874996.1
10
2552
XM_017019508.1
XP_016874997.1
11
2463
XM_017019509.1
XP_016874998.1
9
1472
XM_017019510.1
XP_016874999.1
10
1901
XM_017019511.1
XP_016875000.1
10
2070
XM_017019512.1
XP_016875001.1
10
1426
XM_017019513.1
XP_016875002.1
10
2549
XM_017019514.1
XP_016875003.1
10
2546
XM_017019515.1
XP_016875004.1
10
1892
XM_017019516.1
XP_016875005.1
10
2543
XM_017019517.1
XP_016875006.1
9
2423
XM_017019518.1
XP_016875007.1
9
2417
XM_017019520.1
XP_016875009.1
8
1779
XM_017019521.1
XP_016875010.1
8
1798
XM_017019522.1
XP_016875011.1
9
1276
XM_017019523.1
XP_016875012.1
7
983
XM_017019524.1
XP_016875013.1
9
1374
XM_017019525.1
XP_016875014.1
10
1362
XM_017019526.1
XP_016875015.1
9
1270
XM_017019527.1
XP_016875016.1
9
1289
XM_017019528.1
XP_016875017.1
10
1795
XM_017019529.1
XP_016875018.1
9
1681
XM_017019530.1
XP_016875019.1
8
1586
GenBank mRNA
AK095191.1
-
10
1365
AL832881.1
-
5
907
BC051326.1
-
5
710
6275848
BC074765.2
-
4
450
30915178
L14482.1
-
5
578
Z21966.1
-
4
617
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
heart development
muscle organ development
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-w58vj:80/100.66.78.53:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0