Search
Search

ARL5A| Gene Symbol: ARL5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037174.1 | NP_001032251.1 | 3 | 282 | ||
| NM_012097.3 | NP_036229.1 | 3 | 538 | |||
| NM_177985.2 | NP_817114.2 | 3 | 280 | |||
| GenBank mRNA | AF087889.1 | - | 468 | |||
| AF100740.1 | - | 3 | 285 | |||
| AF493891.1 | - | 3 | 219 | |||
| AK292714.1 | - | 3 | 448 | |||
| AK298937.1 | - | 3 | 226 | |||
| BC001254.1 | - | 3 | 456 | 3453177 | ||
| BG707385.1 | - | 3 | 259 | |||
| BQ216648.1 | - | 3 | 525 | |||
| BT009829.1 | - | 3 | 219 | |||
| BX461356.2 | - | 3 | 253 | |||
| CX867116.1 | - | 3 | 544 | |||