Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FBXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297437.1 | NP_001284366.1 | 1 | 162 | ||
NM_012176.2 | NP_036308.1 | 1 | 160 | |||
NM_033484.2 | NP_277019.1 | 1 | 160 | |||
XM_006714470.3 | XP_006714533.1 | 1 | 94 | |||
XM_011514026.2 | XP_011512328.1 | 1 | 165 | |||
GenBank mRNA | AF129534.1 | - | 1 | 104 | ||
AF176703.1 | - | 1 | 98 | |||
AK001003.1 | - | 1 | 124 | |||
AK024075.1 | - | 1 | 160 | |||
AU143782.1 | - | 1 | 160 | |||
BC040011.1 | - | 1 | 121 | |||
BC048098.1 | - | 1 | 128 | 4830192 |