Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PDX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000209.3 | NP_000200.1 | 2 | 605 | ||
GenBank mRNA | U30329.1 | - | 2 | 517 | ||
U35632.1 | - | 2 | 511 | |||
X99894.1 | - | 2 | 596 |