Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTN SIRNA Assays ›
          Gene Symbol:
          PTN
          Gene Name:
          pleiotrophin
          Gene Aliases:
          HARP, HB-GAM, HBBM, HBGF-8, HBGF8, HBNF, HBNF-1, NEGF1, OSF-1
          Chromosome Location:
          Chr.7: 137227341 - 137343865 on Build GRCh38
          UniGene ID:
          Hs.371249
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 4355

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321386.1 NP_001308315.1 5 608
          NM_001321387.1 NP_001308316.1 5 907
          NM_002825.5 NP_002816.1 5 845
          XM_017012473.1 XP_016867962.1 5 725
          GenBank mRNA AK290488.1 - 5 674
          AK313424.1 - 5 748
          BC005916.1 - 5 833 4248708
          BI462078.1 - 5 619
          BT019692.1 - 4 482
          BX441769.2 - 5 610
          CN413530.1 - 5 594
          CR450338.1 - 4 482
          D90226.1 - 5 606
          M57399.1 - 5 877
          X52946.1 - 5 733

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017066, XM_006236290, XM_006236294, M55601, BC062013, and others.
          This siRNA also targets the following transcripts in Mouse: NM_008973, AK011346, BC002064, EF437379, BC061695, and others.

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          transmembrane receptor protein tyrosine phosphatase signaling pathway
          nervous system development
          heart development
          learning
          positive regulation of cell proliferation
          regulation of cell shape
          positive regulation of cell-substrate adhesion
          positive regulation of neuron projection development
          response to activity
          negative regulation of angiogenesis
          spinal cord development
          cerebellum development
          thalamus development
          bone mineralization
          lung development
          negative regulation of cell migration
          response to estradiol
          response to progesterone
          cellular response to UV
          cellular response to platelet-derived growth factor stimulus
          response to drug
          positive regulation of apoptotic process
          negative regulation of catalytic activity
          estrous cycle
          endothelial cell differentiation
          negative regulation of membrane potential
          negative regulation of epithelial cell proliferation
          positive regulation of cell division
          retinal rod cell differentiation
          negative regulation of glial cell proliferation
          long-term synaptic potentiation
          cellular response to vitamin D
          cellular response to hypoxia
          negative regulation of mesenchymal cell proliferation
          response to kainic acid
          rod bipolar cell differentiation
          response to ciliary neurotrophic factor
          positive regulation of skeletal muscle acetylcholine-gated channel clustering
          negative regulation of neuromuscular junction development
          response to nerve growth factor
          positive regulation of hepatocyte proliferation
          protein phosphatase inhibitor activity
          growth factor activity
          heparin binding
          chondroitin sulfate proteoglycan binding
          chondroitin sulfate binding
          vascular endothelial growth factor binding
          heparan sulfate binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0