Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RNF146 SIRNA Assays ›
          Gene Symbol:
          RNF146
          Gene Name:
          ring finger protein 146
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 127266340 - 127289893 on Build GRCh38
          UniGene ID:
          Hs.267120
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 43636

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RNF146
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242844.1 NP_001229773.1 5 780
          NM_001242845.1 NP_001229774.1 5 629
          NM_001242846.1 NP_001229775.1 5 542
          NM_001242847.1 NP_001229776.1 4 671
          NM_001242848.1 NP_001229777.1 4 401
          NM_001242849.1 NP_001229778.1 3 543
          NM_001242850.1 NP_001229779.1 3 273
          NM_001242851.1 NP_001229780.1 3 213
          NM_001242852.1 NP_001229781.1 5 760
          NM_030963.3 NP_112225.2 5 510
          XM_006715571.3 XP_006715634.1 5 1210
          XM_011536161.2 XP_011534463.1 6 1097
          XM_011536162.2 XP_011534464.1 6 1149
          XM_011536163.2 XP_011534465.1 6 1129
          XM_011536164.2 XP_011534466.1 5 988
          XM_017011336.1 XP_016866825.1 3 890
          XM_017011337.1 XP_016866826.1 5 4131
          XM_017011338.1 XP_016866827.1 7 1238
          XM_017011339.1 XP_016866828.1 6 1267
          XM_017011340.1 XP_016866829.1 6 1319
          XM_017011341.1 XP_016866830.1 4 1018
          XM_017011342.1 XP_016866831.1 5 1159
          XM_017011343.1 XP_016866832.1 6 1268
          GenBank mRNA AJ315122.1 - 1 75
          AJ420549.1 - 2 145
          AK027436.1 - 2 214
          AK027558.1 - 3 262
          AK027776.1 - 1 85
          AL136829.1 - 5 473
          BC008235.1 - 2 162 3868592
          BG708628.1 - 3 508
          BI669435.1 - 5 608
          BI771613.1 - 2 212
          CN388414.1 - 3 353
          CR533514.1 - 1 74
          DA043259.1 - 5 498
          DA145911.1 - 4 388
          DA707096.1 - 4 551
          DB267873.1 - 5 514

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          Wnt signaling pathway
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          protein autoubiquitination
          protein K48-linked ubiquitination
          positive regulation of canonical Wnt signaling pathway
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          ubiquitin protein ligase activity
          poly-ADP-D-ribose binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qq54g:80/100.66.72.153:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0