Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KANK1 SIRNA Assays ›
Gene Symbol:
KANK1
Gene Name:
KN motif and ankyrin repeat domains 1
Gene Aliases:
ANKRD15, CPSQ2, KANK
Chromosome Location:
Chr.9: 470294 - 746106 on Build GRCh38
UniGene ID:
Hs.306764
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44243
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256876.1
NP_001243805.1
7
1238
NM_001256877.1
NP_001243806.1
4
1347
NM_015158.3
NP_055973.2
3
732
NM_153186.4
NP_694856.1
2
994
XM_005251411.2
XP_005251468.1
3
732
XM_005251414.3
XP_005251471.1
2
1195
XM_005251415.2
XP_005251472.1
2
924
XM_005251416.2
XP_005251473.1
2
700
XM_005251417.3
XP_005251474.1
2
1191
XM_005251418.2
XP_005251475.1
2
1202
XM_005251419.2
XP_005251476.1
2
893
XM_006716743.2
XP_006716806.1
3
708
XM_011517819.1
XP_011516121.1
4
823
XM_011517820.1
XP_011516122.1
3
840
XM_011517821.1
XP_011516123.1
3
767
XM_017014511.1
XP_016870000.1
4
945
XM_017014512.1
XP_016870001.1
3
2256
XM_017014513.1
XP_016870002.1
5
1458
XM_017014514.1
XP_016870003.1
3
1144
XM_017014515.1
XP_016870004.1
4
880
XM_017014516.1
XP_016870005.1
3
1144
XM_017014517.1
XP_016870006.1
3
1144
XM_017014518.1
XP_016870007.1
3
732
XM_017014519.1
XP_016870008.1
3
1144
XM_017014520.1
XP_016870009.1
3
732
XM_017014521.1
XP_016870010.1
3
1144
XM_017014522.1
XP_016870011.1
4
945
XM_017014523.1
XP_016870012.1
3
732
XM_017014524.1
XP_016870013.1
3
1144
XM_017014525.1
XP_016870014.1
3
1144
XM_017014526.1
XP_016870015.1
3
1177
XM_017014527.1
XP_016870016.1
2
924
XM_017014528.1
XP_016870017.1
3
778
XM_017014529.1
XP_016870018.1
2
1191
XM_017014530.1
XP_016870019.1
2
700
XM_017014531.1
XP_016870020.1
2
924
XM_017014532.1
XP_016870021.1
2
1198
XM_017014533.1
XP_016870022.1
2
641
XM_017014534.1
XP_016870023.1
2
924
XM_017014535.1
XP_016870024.1
2
1190
XM_017014536.1
XP_016870025.1
2
893
XM_017014537.1
XP_016870026.1
2
713
XM_017014538.1
XP_016870027.1
2
1198
XM_017014539.1
XP_016870028.1
2
713
XM_017014540.1
XP_016870029.1
2
1189
XM_017014541.1
XP_016870030.1
2
1193
XM_017014542.1
XP_016870031.1
2
700
GenBank mRNA
AK026421.1
-
1
345
AK290028.1
-
3
698
AK292989.1
-
3
711
AK311119.1
-
2
933
AL832454.2
-
3
724
AL833161.1
-
3
712
BC037495.2
-
7
1241
5265517
BC038116.2
-
2
904
5261060
BX648931.1
-
3
726
D79994.2
-
3
1297
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of neuron projection development
positive regulation of Wnt signaling pathway
negative regulation of cell migration
negative regulation of actin filament polymerization
negative regulation of Rho protein signal transduction
positive regulation of catenin import into nucleus
negative regulation of insulin receptor signaling pathway
positive regulation of wound healing
negative regulation of substrate adhesion-dependent cell spreading
negative regulation of ruffle assembly
regulation of establishment of cell polarity
negative regulation of lamellipodium morphogenesis
protein binding
beta-catenin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0