Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: WAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016628.4 | NP_057712.2 | 2, 3 | 165 | ||
NM_100264.2 | NP_567822.1 | 2, 3 | 456 | |||
NM_100486.3 | NP_567823.1 | 2, 3 | 165 | |||
XM_017016315.1 | XP_016871804.1 | 2, 3 | 276 | |||
XM_017016317.1 | XP_016871806.1 | 2, 3 | 276 | |||
XM_017016318.1 | XP_016871807.1 | 2, 3 | 276 | |||
GenBank mRNA | AK055852.1 | - | 2, 3 | 273 | ||
AK223600.1 | - | 2, 3 | 163 | |||
AK225846.1 | - | 2, 3 | 269 | |||
AK290174.1 | - | 2, 3 | 170 | |||
BC004258.1 | - | 2, 3 | 156 | 3347345 | ||
BI226998.1 | - | 2, 3 | 53 | |||
BI463901.1 | - | 2, 3 | 556 | |||
CB143386.1 | - | 2, 3 | 344 |