Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KANK1 SIRNA Assays ›
Gene Symbol:
KANK1
Gene Name:
KN motif and ankyrin repeat domains 1
Gene Aliases:
ANKRD15, CPSQ2, KANK
Chromosome Location:
Chr.9: 470294 - 746106 on Build GRCh38
UniGene ID:
Hs.306764
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44336
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256876.1
NP_001243805.1
7
1284
NM_001256877.1
NP_001243806.1
4
1393
NM_015158.3
NP_055973.2
3
778
NM_153186.4
NP_694856.1
2
1040
XM_005251411.2
XP_005251468.1
3
778
XM_005251414.3
XP_005251471.1
2
1241
XM_005251415.2
XP_005251472.1
2
970
XM_005251416.2
XP_005251473.1
2
746
XM_005251417.3
XP_005251474.1
2
1237
XM_005251418.2
XP_005251475.1
2
1248
XM_005251419.2
XP_005251476.1
2
939
XM_006716743.2
XP_006716806.1
3
754
XM_011517819.1
XP_011516121.1
4
869
XM_011517820.1
XP_011516122.1
3
886
XM_011517821.1
XP_011516123.1
3
813
XM_017014511.1
XP_016870000.1
4
991
XM_017014512.1
XP_016870001.1
3
2302
XM_017014513.1
XP_016870002.1
5
1504
XM_017014514.1
XP_016870003.1
3
1190
XM_017014515.1
XP_016870004.1
4
926
XM_017014516.1
XP_016870005.1
3
1190
XM_017014517.1
XP_016870006.1
3
1190
XM_017014518.1
XP_016870007.1
3
778
XM_017014519.1
XP_016870008.1
3
1190
XM_017014520.1
XP_016870009.1
3
778
XM_017014521.1
XP_016870010.1
3
1190
XM_017014522.1
XP_016870011.1
4
991
XM_017014523.1
XP_016870012.1
3
778
XM_017014524.1
XP_016870013.1
3
1190
XM_017014525.1
XP_016870014.1
3
1190
XM_017014526.1
XP_016870015.1
3
1223
XM_017014527.1
XP_016870016.1
2
970
XM_017014528.1
XP_016870017.1
3
824
XM_017014529.1
XP_016870018.1
2
1237
XM_017014530.1
XP_016870019.1
2
746
XM_017014531.1
XP_016870020.1
2
970
XM_017014532.1
XP_016870021.1
2
1244
XM_017014533.1
XP_016870022.1
2
687
XM_017014534.1
XP_016870023.1
2
970
XM_017014535.1
XP_016870024.1
2
1236
XM_017014536.1
XP_016870025.1
2
939
XM_017014537.1
XP_016870026.1
2
759
XM_017014538.1
XP_016870027.1
2
1244
XM_017014539.1
XP_016870028.1
2
759
XM_017014540.1
XP_016870029.1
2
1235
XM_017014541.1
XP_016870030.1
2
1239
XM_017014542.1
XP_016870031.1
2
746
GenBank mRNA
AK026421.1
-
1
391
AK290028.1
-
3
744
AK292989.1
-
3
757
AL832454.2
-
3
770
AL833161.1
-
3
758
BC037495.2
-
7
1287
5265517
BC038116.2
-
2
950
5261060
BX648931.1
-
3
772
D79994.2
-
3
1343
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of neuron projection development
positive regulation of Wnt signaling pathway
negative regulation of cell migration
negative regulation of actin filament polymerization
negative regulation of Rho protein signal transduction
positive regulation of catenin import into nucleus
negative regulation of insulin receptor signaling pathway
positive regulation of wound healing
negative regulation of substrate adhesion-dependent cell spreading
negative regulation of ruffle assembly
regulation of establishment of cell polarity
negative regulation of lamellipodium morphogenesis
protein binding
beta-catenin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-57bf8:80/100.66.79.204:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0