Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IFFO1 SIRNA Assays ›
          Gene Symbol:
          IFFO1
          Gene Name:
          intermediate filament family orphan 1
          Gene Aliases:
          HOM-TES-103, IFFO
          Chromosome Location:
          Chr.12: 6539528 - 6556102 on Build GRCh38
          UniGene ID:
          Hs.15243
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 44500

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IFFO1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039670.2 NP_001034759.1 6, 7 1298
          NM_001193457.1 NP_001180386.1 6, 7 1322
          NM_080730.4 NP_542768.2 6, 7 1298
          NR_036467.1 6, 7 1395
          XM_006718975.3 XP_006719038.1 6, 7 1314
          XM_006718976.3 XP_006719039.1 5, 6 1308
          XM_006718977.3 XP_006719040.1 5, 6 1308
          XM_006718981.3 XP_006719044.1 7, 8 983
          XM_011520947.2 XP_011519249.1 6, 7 1317
          XM_011520948.2 XP_011519250.1 6, 7 1314
          XM_011520949.2 XP_011519251.1 5, 6 1308
          XM_011520952.2 XP_011519254.1 7, 8 983
          XM_017019169.1 XP_016874658.1 6, 7 1317
          XM_017019170.1 XP_016874659.1 6, 7 1120
          XM_017019171.1 XP_016874660.1 6, 7 1314
          XM_017019172.1 XP_016874661.1 7, 8 986
          XM_017019173.1 XP_016874662.1 6, 7 617
          GenBank mRNA AF124432.1 - 6, 7 959
          AK000710.1 - 1, 2 531
          AK095106.1 - 2, 3 876
          AK298905.1 - 6, 7 531
          BC001790.1 - 6, 7 1375
          BC002857.2 - 5, 6 436
          BC004384.1 - 5, 6 436
          BC010431.1 - 6, 7 1375
          BC023664.2 - 6, 7 1375
          BC063004.1 - 6, 7 573
          BC071170.1 - 1, 2 447
          BC103817.1 - 1, 2 452
          BC103818.1 - 1, 2 452
          BC110387.1 - 6, 7 1288 5183989
          BC113848.1 - 1, 2 452
          BC114454.1 - 1, 2 452
          BM922290.1 - 471
          BX327856.2 - 6, 7 514
          BX648701.1 - 7, 8 884
          BX648743.1 - 7, 8 884
          DA268043.1 - 1, 2 513

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001108647, XM_006237373, XM_006237372, XM_008763292, XM_006237376, and others.
          This siRNA also targets the following transcripts in Mouse: AK044709, AK028273, AK189508, BC057601, AK031129, and others.

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Process(es)

          structural molecule activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0