Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other YY1AP1 SIRNA Assays ›
Gene Symbol:
YY1AP1
Gene Name:
YY1 associated protein 1
Gene Aliases:
HCCA1, HCCA2, YAP, YY1AP
Chromosome Location:
Chr.1: 155659442 - 155689032 on Build GRCh38
UniGene ID:
Hs.584927
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44511
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: YY1AP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198899.1
NP_001185828.1
1
394
NM_001198900.1
NP_001185829.1
3
327
NM_001198901.1
NP_001185830.1
3
217
NM_001198902.1
NP_001185831.1
3
184
NM_001198903.1
NP_001185832.1
3
704
NM_001198904.1
NP_001185833.1
3
704
NM_001198905.1
NP_001185834.1
4
368
NM_001198906.1
NP_001185835.1
4
352
NM_018253.3
NP_060723.2
5
449
NM_139118.2
NP_620829.1
4
352
NM_139119.2
NP_620830.1
4
365
NM_139121.2
NP_620832.1
3
162
GenBank mRNA
AF203474.1
-
3
384
AF206328.1
-
4
284
AF466401.1
-
3
158
AK023976.1
-
4
320
AK297562.1
-
3
517
AK298904.1
-
4
348
AK303046.1
-
3
367
AK303386.1
-
1
372
AK309701.1
-
3
523
AK315049.1
-
4
399
AY604179.1
-
3
189
BC001655.2
-
5
379
2961648
BC001843.2
-
5
379
2961648
BC003500.1
-
3
162
BC008766.2
-
3
261
3051391
BC014906.2
-
4
297
3922542
BC044887.1
-
3
324
5276742
BG325066.1
-
3
185
CR749789.1
-
4
339
DC300265.1
-
3
217
DC346197.1
-
1
394
DC388975.1
-
4
353
DC399389.1
-
4
368
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of cell cycle
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0