Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKS1B SIRNA Assays ›
          Gene Symbol:
          ANKS1B
          Gene Name:
          ankyrin repeat and sterile alpha motif domain containing 1B
          Gene Aliases:
          AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
          Chromosome Location:
          Chr.12: 98729904 - 99984727 on Build GRCh38
          UniGene ID:
          Hs.506458
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 44520

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKS1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204066.1 NP_001190995.1 3, 4 262
          NM_001204067.1 NP_001190996.1 3, 4 323
          NM_001204068.1 NP_001190997.1 2, 3 606
          NM_001204069.1 NP_001190998.1 2, 3 606
          NM_001204070.1 NP_001190999.1 2, 3 606
          NM_020140.3 NP_064525.1 2, 3 606
          NM_152788.4 NP_690001.3 16, 17 3038
          NM_181670.3 NP_858056.2 2, 3 606
          XM_005269028.4 XP_005269085.1 16, 17 2649
          XM_005269029.4 XP_005269086.1 16, 17 2609
          XM_006719504.3 XP_006719567.1 16, 17 3111
          XM_006719505.3 XP_006719568.1 16, 17 3111
          XM_006719506.3 XP_006719569.1 16, 17 3111
          XM_006719507.3 XP_006719570.1 16, 17 3111
          XM_006719508.3 XP_006719571.1 16, 17 3111
          XM_006719509.3 XP_006719572.1 16, 17 3111
          XM_006719510.3 XP_006719573.1 16, 17 3111
          XM_006719512.3 XP_006719575.1 16, 17 3111
          XM_006719513.3 XP_006719576.1 16, 17 3111
          XM_006719514.3 XP_006719577.1 16, 17 3099
          XM_011538571.2 XP_011536873.1 16, 17 3111
          XM_017019651.1 XP_016875140.1 16, 17 3111
          XM_017019652.1 XP_016875141.1 16, 17 3099
          XM_017019653.1 XP_016875142.1 16, 17 3111
          XM_017019654.1 XP_016875143.1 16, 17 3099
          XM_017019655.1 XP_016875144.1 16, 17 3111
          XM_017019656.1 XP_016875145.1 16, 17 3111
          XM_017019657.1 XP_016875146.1 16, 17 3111
          XM_017019658.1 XP_016875147.1 16, 17 3111
          XM_017019659.1 XP_016875148.1 2, 3 602
          XM_017019660.1 XP_016875149.1 2, 3 602
          XM_017019661.1 XP_016875150.1 3, 4 316
          XM_017019662.1 XP_016875151.1 3, 4 446
          XM_017019663.1 XP_016875152.1 2, 3 602
          XM_017019664.1 XP_016875153.1 3, 4 258
          XM_017019665.1 XP_016875154.1 2, 3 602
          GenBank mRNA AF145204.1 - 10, 11 1742
          AF164792.1 - 2, 3 539
          AK289768.1 - 2, 3 571
          AK294191.1 - 3, 4 323
          AY281131.1 - 16, 17 2624
          AY281132.1 - 2, 3 299
          AY283057.2 - 9, 10 1655
          AY620824.1 - 7, 8 1349
          AY753193.1 - 2, 3 299
          BC026313.1 - 2, 3 501
          BC068451.1 - 3, 4 262
          BC142669.1 - 2, 3 553
          BC150204.1 - 2, 3 553

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of synaptic plasticity by receptor localization to synapse
          ephrin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0