Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKS1B SIRNA Assays ›
Gene Symbol:
ANKS1B
Gene Name:
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases:
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
Chromosome Location:
Chr.12: 98729904 - 99984727 on Build GRCh38
UniGene ID:
Hs.506458
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44587
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKS1B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204066.1
NP_001190995.1
3, 4
274
NM_001204067.1
NP_001190996.1
3, 4
335
NM_001204068.1
NP_001190997.1
2, 3
618
NM_001204069.1
NP_001190998.1
2, 3
618
NM_001204070.1
NP_001190999.1
2, 3
618
NM_020140.3
NP_064525.1
2, 3
618
NM_152788.4
NP_690001.3
16, 17
3050
NM_181670.3
NP_858056.2
2, 3
618
XM_005269028.4
XP_005269085.1
16, 17
2661
XM_005269029.4
XP_005269086.1
16, 17
2621
XM_006719504.3
XP_006719567.1
16, 17
3123
XM_006719505.3
XP_006719568.1
16, 17
3123
XM_006719506.3
XP_006719569.1
16, 17
3123
XM_006719507.3
XP_006719570.1
16, 17
3123
XM_006719508.3
XP_006719571.1
16, 17
3123
XM_006719509.3
XP_006719572.1
16, 17
3123
XM_006719510.3
XP_006719573.1
16, 17
3123
XM_006719512.3
XP_006719575.1
16, 17
3123
XM_006719513.3
XP_006719576.1
16, 17
3123
XM_006719514.3
XP_006719577.1
16, 17
3111
XM_011538571.2
XP_011536873.1
16, 17
3123
XM_017019651.1
XP_016875140.1
16, 17
3123
XM_017019652.1
XP_016875141.1
16, 17
3111
XM_017019653.1
XP_016875142.1
16, 17
3123
XM_017019654.1
XP_016875143.1
16, 17
3111
XM_017019655.1
XP_016875144.1
16, 17
3123
XM_017019656.1
XP_016875145.1
16, 17
3123
XM_017019657.1
XP_016875146.1
16, 17
3123
XM_017019658.1
XP_016875147.1
16, 17
3123
XM_017019659.1
XP_016875148.1
2, 3
614
XM_017019660.1
XP_016875149.1
2, 3
614
XM_017019661.1
XP_016875150.1
3, 4
328
XM_017019662.1
XP_016875151.1
3, 4
458
XM_017019663.1
XP_016875152.1
2, 3
614
XM_017019664.1
XP_016875153.1
3, 4
270
XM_017019665.1
XP_016875154.1
2, 3
614
GenBank mRNA
AF145204.1
-
10, 11
1754
AF164792.1
-
2, 3
551
AK289768.1
-
2, 3
583
AK294191.1
-
3, 4
335
AY281131.1
-
16, 17
2636
AY281132.1
-
2, 3
311
AY283057.2
-
9, 10
1667
AY620824.1
-
7, 8
1361
AY753193.1
-
2, 3
311
BC026313.1
-
2, 3
513
BC068451.1
-
3, 4
274
BC142669.1
-
2, 3
565
BC150204.1
-
2, 3
565
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of synaptic plasticity by receptor localization to synapse
ephrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0