Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TIRAP SIRNA Assays ›
          Gene Symbol:
          TIRAP
          Gene Name:
          TIR domain containing adaptor protein
          Gene Aliases:
          BACTS1, Mal, MyD88-2, wyatt
          Chromosome Location:
          Chr.11: 126281268 - 126294933 on Build GRCh38
          UniGene ID:
          Hs.537126
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 44728

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TIRAP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039661.1 NP_001034750.1 5 1014
          NM_001318776.1 NP_001305705.1 4 861
          NM_001318777.1 NP_001305706.1 4 861
          NM_148910.2 NP_683708.1 5 1030
          XM_005271399.3 XP_005271456.2 3 759
          XM_011542576.2 XP_011540878.1 3 759
          XM_017017161.1 XP_016872650.1 2 2382
          XM_017017162.1 XP_016872651.1 5 1051
          GenBank mRNA AB446477.1 - 2 578
          AF378129.1 - 5 1011
          AF406652.1 - 2 578
          AF410783.1 - 5 1007
          AK124298.1 - 4 861
          AK313147.1 - 4 854
          AY576785.1 - 2 617
          AY576786.1 - 2 617
          AY576787.1 - 2 617
          BC032474.1 - 5 983 5216171

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MyD88-dependent toll-like receptor signaling pathway
          inflammatory response
          cell surface receptor signaling pathway
          activation of NF-kappaB-inducing kinase activity
          myeloid cell differentiation
          positive regulation of B cell proliferation
          response to lipopolysaccharide
          regulation of interferon-beta production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-15 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          positive regulation of toll-like receptor 2 signaling pathway
          positive regulation of toll-like receptor 3 signaling pathway
          toll-like receptor 4 signaling pathway
          positive regulation of toll-like receptor 4 signaling pathway
          TIRAP-dependent toll-like receptor 4 signaling pathway
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of growth of symbiont in host
          innate immune response
          regulation of innate immune response
          positive regulation of interleukin-6 biosynthetic process
          positive regulation of JNK cascade
          defense response to Gram-positive bacterium
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of ERK1 and ERK2 cascade
          3'-UTR-mediated mRNA stabilization
          cellular response to bacterial lipopeptide
          cellular response to lipoteichoic acid
          positive regulation of neutrophil chemotaxis
          positive regulation of protein homodimerization activity
          positive regulation of chemokine (C-X-C motif) ligand 1 production
          positive regulation of chemokine (C-X-C motif) ligand 2 production
          protein kinase C binding
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          protein binding, bridging
          Toll-like receptor 4 binding
          Toll-like receptor 2 binding
          protein homodimerization activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0