Search
Search

MAGEA4| Gene Symbol: MAGEA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011548.1 | NP_001011548.1 | 3 | 535 | ||
| NM_001011549.1 | NP_001011549.1 | 3 | 532 | |||
| NM_001011550.1 | NP_001011550.1 | 3 | 523 | |||
| NM_002362.4 | NP_002353.3 | 3 | 538 | |||
| XM_005274677.2 | XP_005274734.1 | 3 | 578 | |||
| XM_005274678.3 | XP_005274735.1 | 3 | 536 | |||
| XM_005274679.3 | XP_005274736.1 | 3 | 545 | |||
| GenBank mRNA | AK300179.1 | - | 2 | 595 | ||
| AK313862.1 | - | 3 | 574 | |||
| AY954624.1 | - | 2 | 408 | |||
| BC017723.1 | - | 3 | 535 | 4394783 | ||
| BC096712.1 | - | 3 | 474 | |||
| BM548926.1 | - | 3 | 532 | |||
| BM562570.1 | - | 2 | 613 | |||
| BX479311.1 | - | 3 | 535 | |||
| D32075.1 | - | 1 | 382 | |||
| D32077.1 | - | 1 | 382 | |||
| U10340.1 | - | 1 | 317 | |||