Search
Search

SOD3| Gene Symbol: SOD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003102.2 | NP_003093.2 | 2 | 312 | ||
| GenBank mRNA | AK313935.1 | - | 2 | 293 | ||
| BC014418.1 | - | 2 | 177 | 4644224 | ||
| CR541853.1 | - | 1 | 107 | |||
| DA668533.1 | - | 2 | 297 | |||
| J02947.1 | - | 2 | 176 | |||