Search
Search

GNG5| Gene Symbol: GNG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005274.2 | NP_005265.1 | 3 | 462 | ||
| GenBank mRNA | AF038955.1 | - | 2 | 145 | ||
| AF493873.1 | - | 2 | 108 | |||
| AK312111.1 | - | 2 | 221 | |||
| BC003563.1 | - | 2 | 191 | 3502879 | ||
| BM809260.1 | - | 2 | 201 | |||
| BT006823.1 | - | 2 | 108 | |||