Search
Search

GPR12| Gene Symbol: GPR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005288.3 | NP_005279.1 | 2 | 619 | ||
| GenBank mRNA | AK290220.1 | - | 2 | 620 | ||
| AK307756.1 | - | 2 | 619 | |||
| AK312602.1 | - | 2 | 475 | |||
| BC067448.1 | - | 1 | 397 | |||
| BC067449.1 | - | 1 | 397 | |||
| BC067450.1 | - | 1 | 397 | |||
| BC067451.1 | - | 1 | 397 | |||
| BC067452.1 | - | 1 | 397 | |||
| BC112144.1 | - | 1 | 472 | 8327612 | ||
| BC112146.1 | - | 1 | 472 | 8327614 | ||
| HQ995525.1 | - | 1 | 397 | |||
Set Membership: |
|