Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF7 SIRNA Assays ›
Gene Symbol:
TCF7
Gene Name:
transcription factor 7 (T-cell specific, HMG-box)
Gene Aliases:
TCF-1
Chromosome Location:
Chr.5: 134114703 - 134148229 on Build GRCh38
UniGene ID:
Hs.573153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4513
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134851.2
NP_001128323.2
8
809
NM_003202.3
NP_003193.2
9
1253
NM_201632.3
NP_963963.1
8
809
NM_201634.3
NP_963965.1
8
809
NM_213648.3
NP_998813.1
7
1043
NR_033449.1
7
1044
XM_006714678.3
XP_006714741.1
10
1354
XM_006714679.3
XP_006714742.1
9
1261
XM_006714680.3
XP_006714743.1
10
1354
XM_006714682.2
XP_006714745.1
10
1354
XM_006714684.2
XP_006714747.1
9
1261
XM_006714685.3
XP_006714748.1
9
899
XM_006714686.3
XP_006714749.1
8
808
XM_006714687.3
XP_006714750.1
8
766
XM_011543604.2
XP_011541906.1
10
1354
XM_011543606.1
XP_011541908.1
10
1354
XM_011543607.2
XP_011541909.1
8
6842
XM_011543608.2
XP_011541910.1
8
2887
XM_011543609.2
XP_011541911.1
7
755
XM_011543610.2
XP_011541912.1
8
887
XM_011543611.2
XP_011541913.1
8
1781
XM_011543612.2
XP_011541914.1
7
755
XM_011543613.2
XP_011541915.1
7
794
XM_017009789.1
XP_016865278.1
9
899
XM_017009790.1
XP_016865279.1
8
1781
GenBank mRNA
AK057580.1
-
7
1044
AK093530.1
-
7
914
AK093683.1
-
4
1143
AK131428.1
-
4
1262
AK310591.1
-
7
755
AL832816.1
-
9
969
AL834166.1
-
8
979
BC048769.1
-
9
1253
6063270
BC072023.1
-
9
1188
6292932
X59869.1
-
8
791
X59870.1
-
8
791
X59871.1
-
8
791
Z47361.1
-
7
713
Z47362.1
-
7
713
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
neural tube development
embryonic genitalia morphogenesis
T cell receptor V(D)J recombination
regulation of cell proliferation
canonical Wnt signaling pathway involved in negative regulation of apoptotic process
alpha-beta T cell differentiation
embryonic digestive tract morphogenesis
embryonic hindgut morphogenesis
canonical Wnt signaling pathway
cellular response to interleukin-4
beta-catenin-TCF complex assembly
core promoter binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein binding
beta-catenin binding
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-wf5dr:80/100.66.77.5:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0