Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PBDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300888.1 | NP_001287817.1 | 2 | 297 | ||
NM_016500.3 | NP_057584.2 | 2 | 290 | |||
GenBank mRNA | BC001220.1 | - | 2 | 290 | 3528586 | |
BC051894.1 | - | 2 | 152 | 6528202 | ||
BM817702.1 | - | 2 | 186 | |||
CD704296.1 | - | 2 | 191 |