Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAC1 SIRNA Assays ›
          Gene Symbol:
          RAC1
          Gene Name:
          ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1)
          Gene Aliases:
          MIG5, Rac-1, TC-25, p21-Rac1
          Chromosome Location:
          Chr.7: 6374495 - 6403967 on Build GRCh38
          UniGene ID:
          Hs.413812
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 45358

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006908.4 NP_008839.2 2, 3 340
          NM_018890.3 NP_061485.1 2, 3 340
          GenBank mRNA AB209410.1 - 2, 3 287
          AB451238.1 - 2, 3 99
          AB451363.1 - 2, 3 99
          AF136373.1 - 2, 3 99
          AF498964.1 - 2, 3 99
          AJ132694.1 - 2, 3 99
          AK054993.1 - 2, 3 315
          AK291806.1 - 2, 3 296
          AK291815.1 - 2, 3 286
          AY279384.1 - 2, 3 257
          BC004247.1 - 2, 3 237 3610745
          BC050687.1 - 2, 3 287 6149377
          BC107748.1 - 2, 3 302 6699945
          BG437715.1 - 2, 3 339
          BQ689587.1 - 3, 4 356
          BT007121.1 - 2, 3 99
          M29870.1 - 2, 3 99
          M31467.1 - 2, 3 99

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          auditory receptor cell morphogenesis
          mast cell chemotaxis
          epithelial cell morphogenesis
          movement of cell or subcellular component
          inflammatory response
          hyperosmotic response
          cell adhesion
          cell-matrix adhesion
          G-protein coupled receptor signaling pathway
          small GTPase mediated signal transduction
          axon guidance
          blood coagulation
          cell proliferation
          regulation of cell size
          response to wounding
          anatomical structure morphogenesis
          regulation of hydrogen peroxide metabolic process
          positive regulation of lamellipodium assembly
          regulation of fibroblast migration
          positive regulation of cell-substrate adhesion
          regulation of neuron maturation
          cerebral cortex radially oriented cell migration
          embryonic olfactory bulb interneuron precursor migration
          cerebral cortex GABAergic interneuron development
          lamellipodium assembly
          actin cytoskeleton organization
          actin filament polymerization
          platelet activation
          regulation of cell migration
          positive regulation of actin filament polymerization
          T cell costimulation
          ruffle organization
          negative regulation of interleukin-23 production
          substrate adhesion-dependent cell spreading
          positive regulation of Rho protein signal transduction
          intracellular signal transduction
          Fc-epsilon receptor signaling pathway
          Fc-gamma receptor signaling pathway involved in phagocytosis
          positive regulation of apoptotic process
          positive regulation of phosphatidylinositol 3-kinase activity
          engulfment of apoptotic cell
          cell-cell junction organization
          bone resorption
          positive regulation of DNA replication
          vascular endothelial growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          negative regulation of receptor-mediated endocytosis
          anatomical structure arrangement
          dendrite morphogenesis
          cell motility
          homeostasis of number of cells within a tissue
          regulation of defense response to virus by virus
          regulation of small GTPase mediated signal transduction
          positive regulation of stress fiber assembly
          localization within membrane
          positive regulation of focal adhesion assembly
          synaptic transmission, GABAergic
          Wnt signaling pathway, planar cell polarity pathway
          regulation of respiratory burst
          cellular response to mechanical stimulus
          semaphorin-plexin signaling pathway
          dopaminergic neuron differentiation
          protein localization to plasma membrane
          positive regulation of neutrophil chemotaxis
          cochlea morphogenesis
          ruffle assembly
          positive regulation of substrate adhesion-dependent cell spreading
          GTPase activity
          protein binding
          GTP binding
          Rab GTPase binding
          enzyme binding
          protein kinase binding
          GTP-dependent protein binding
          thioesterase binding
          histone deacetylase binding
          Rho GDP-dissociation inhibitor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9c8cf:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0