Search
Search

HIRA| Gene Symbol: HIRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003325.3 | NP_003316.3 | 3 | 333 | ||
| GenBank mRNA | AK289809.1 | - | 3 | 113 | ||
| AK299950.1 | - | 4 | 272 | |||
| AK302835.1 | - | 4 | 377 | |||
| AK308642.1 | - | 4 | 272 | |||
| BC032721.1 | - | 3 | 371 | |||
| BC039835.1 | - | 3 | 328 | 5491361 | ||
| CR456503.1 | - | 3 | 119 | |||
| X75296.1 | - | 3 | 340 | |||
| X77633.1 | - | 2 | 89 | |||
| X81844.1 | - | 3 | 174 | |||
| X89887.1 | - | 3 | 333 | |||