Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGF8 SIRNA Assays ›
          Gene Symbol:
          FGF8
          Gene Name:
          fibroblast growth factor 8
          Gene Aliases:
          AIGF, FGF-8, HBGF-8, HH6, KAL6
          Chromosome Location:
          Chr.10: 101770130 - 101780369 on Build GRCh38
          UniGene ID:
          Hs.57710
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 45555

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FGF8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001206389.1 NP_001193318.1 4, 5 362
          NM_006119.4 NP_006110.1 4, 5 455
          NM_033163.3 NP_149353.1 5, 6 542
          NM_033164.3 NP_149354.1 5, 6 509
          NM_033165.3 NP_149355.1 4, 5 422
          GenBank mRNA AB014615.1 - 5, 6 449
          BC069106.1 - 5, 6 454 7216922
          BC128235.1 - 4, 5 422 40114671
          BC128236.1 - 4, 5 422 40114672
          CN369230.1 - 4, 5 372
          U36223.1 - 4, 5 362
          U46211.1 - 4, 5 320
          U46212.1 - 4, 5 353
          U46213.1 - 5, 6 407
          U56978.1 - 3, 4 325

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          patterning of blood vessels
          metanephros development
          branching involved in ureteric bud morphogenesis
          organ induction
          mesonephros development
          neural plate morphogenesis
          heart looping
          blood vessel remodeling
          outflow tract septum morphogenesis
          epithelial to mesenchymal transition involved in endocardial cushion formation
          apoptotic process
          response to oxidative stress
          gastrulation
          motor neuron axon guidance
          mesodermal cell migration
          positive regulation of cell proliferation
          gonad development
          fibroblast growth factor receptor signaling pathway
          anatomical structure morphogenesis
          positive regulation of gene expression
          regulation of phosphatidylinositol 3-kinase signaling
          response to organic cyclic compound
          peptidyl-tyrosine phosphorylation
          pallium development
          subpallium development
          forebrain dorsal/ventral pattern formation
          cell proliferation in forebrain
          forebrain neuron development
          signal transduction involved in regulation of gene expression
          BMP signaling pathway
          male genitalia development
          thyroid gland development
          otic vesicle formation
          midbrain-hindbrain boundary development
          dorsal/ventral axon guidance
          embryonic hindlimb morphogenesis
          aorta morphogenesis
          phosphatidylinositol-3-phosphate biosynthetic process
          odontogenesis
          regulation of odontogenesis of dentin-containing tooth
          response to drug
          negative regulation of neuron apoptotic process
          positive regulation of GTPase activity
          cell fate commitment
          positive regulation of mitotic nuclear division
          positive regulation of organ growth
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          forebrain morphogenesis
          positive chemotaxis
          positive regulation of cell division
          negative regulation of cardiac muscle tissue development
          pharyngeal system development
          canonical Wnt signaling pathway
          corticotropin hormone secreting cell differentiation
          thyroid-stimulating hormone-secreting cell differentiation
          bone development
          lung morphogenesis
          branching involved in salivary gland morphogenesis
          neuroepithelial cell differentiation
          positive regulation of ERK1 and ERK2 cascade
          dopaminergic neuron differentiation
          cell migration involved in mesendoderm migration
          protein tyrosine kinase activity
          Ras guanyl-nucleotide exchange factor activity
          fibroblast growth factor receptor binding
          growth factor activity
          1-phosphatidylinositol-3-kinase activity
          chemoattractant activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0