Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALDOA SIRNA Assays ›
          Gene Symbol:
          ALDOA
          Gene Name:
          aldolase, fructose-bisphosphate A
          Gene Aliases:
          ALDA, GSD12, HEL-S-87p
          Chromosome Location:
          Chr.16: 30053090 - 30070420 on Build GRCh38
          UniGene ID:
          Hs.513490
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 45916

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ALDOA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000034.3 NP_000025.1 8 1384
          NM_001127617.2 NP_001121089.1 3 606
          NM_001243177.1 NP_001230106.1 4 727
          NM_184041.2 NP_908930.1 3 573
          NM_184043.2 NP_908932.1 3 545
          XM_011545768.2 XP_011544070.1 5 1247
          GenBank mRNA AK026577.1 - 3 423
          AK098778.1 - 6 1425
          AK301993.1 - 4 539
          BC004333.1 - 8 1304 3630974
          BC010660.1 - 3 491 3869603
          BC012880.1 - 3 419 3459009
          BC013614.1 - 3 502 3880857
          BC015888.2 - 3 415 3911623
          BC016800.1 - 4 443 4072487
          BQ719865.1 - 3 534
          BX647566.1 - 3 395
          CB151262.1 - 3 544
          CR536528.1 - 2 297
          CR541880.1 - 2 297
          DA564075.1 - 3 543
          DA893541.1 - 3 453
          FJ474908.1 - 4 464
          M11560.1 - 4 464

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          aldolase

          Gene Ontology Categories:

          Function(s) Process(es)

          platelet degranulation
          fructose metabolic process
          gluconeogenesis
          glycolytic process
          ATP biosynthetic process
          striated muscle contraction
          actin filament organization
          regulation of cell shape
          fructose 1,6-bisphosphate metabolic process
          muscle cell cellular homeostasis
          protein homotetramerization
          canonical glycolysis
          cell-cell adhesion
          actin binding
          fructose-bisphosphate aldolase activity
          protein binding
          cytoskeletal protein binding
          tubulin binding
          identical protein binding
          poly(A) RNA binding
          fructose binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-6pmgv:80/100.66.77.221:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0