Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCL19 SIRNA Assays ›
          Gene Symbol:
          CCL19
          Gene Name:
          C-C motif chemokine ligand 19
          Gene Aliases:
          CKb11, ELC, MIP-3b, MIP3B, SCYA19
          Chromosome Location:
          Chr.9: 34689570 - 34691277 on Build GRCh38
          UniGene ID:
          Hs.50002
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 46152

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCL19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006274.2 NP_006265.1 2 228
          GenBank mRNA AB000887.1 - 2 228
          AK311945.1 - 2 227
          AX772806.1 - 2 227
          BC027968.1 - 2 214 5225849
          CR456868.1 - 2 90
          U77180.1 - 2 90
          U88321.1 - 2 209

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001108661.
          This siRNA also targets the following transcripts in Mouse: NM_011888, AF059208, BC051472, AK156269, BC025130, and others.

          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of T cell polarity
          immunological synapse formation
          dendritic cell chemotaxis
          myeloid dendritic cell chemotaxis
          monocyte chemotaxis
          positive regulation of dendritic cell antigen processing and presentation
          cellular calcium ion homeostasis
          inflammatory response
          immune response
          cell communication
          G-protein coupled receptor signaling pathway
          activation of JUN kinase activity
          response to virus
          positive regulation of glycoprotein biosynthetic process
          T cell costimulation
          positive regulation of interleukin-12 production
          positive regulation of tumor necrosis factor production
          response to prostaglandin E
          positive regulation of T cell proliferation
          positive regulation of NF-kappaB import into nucleus
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of GTPase activity
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of T-helper 1 cell differentiation
          positive regulation of endocytosis
          positive regulation of protein kinase activity
          positive regulation of JNK cascade
          lymphocyte chemotaxis
          positive regulation of receptor-mediated endocytosis
          cell maturation
          positive regulation of interleukin-1 beta secretion
          positive regulation of chemotaxis
          release of sequestered calcium ion into cytosol
          positive regulation of protein kinase B signaling
          regulation of cell projection assembly
          chemokine-mediated signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          response to nitric oxide
          interleukin-12 secretion
          positive regulation of neutrophil chemotaxis
          mature conventional dendritic cell differentiation
          cellular response to virus
          negative regulation of leukocyte apoptotic process
          positive regulation of cell motility
          positive regulation of dendritic cell dendrite assembly
          chemokine activity
          CCR7 chemokine receptor binding
          CCR10 chemokine receptor binding
          chemokine receptor binding
          CCR chemokine receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0