Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD24 SIRNA Assays ›
          Gene Symbol:
          Cd24
          Gene Name:
          CD24 molecule
          Gene Aliases:
          Cd24a
          Chromosome Location:
          Chr.20: 48335540 - 48340847 on Build Rnor_6.0
          UniGene ID:
          Rn.6007
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 46590

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cd24
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012752.3 NP_036884.1 2 316
          GenBank mRNA BC064439.1 - 2 300 5623514
          BC081851.1 - 2 311 7110912
          DY315580.1 - 2 345
          U49062.1 - 2 300
          Z11663.1 - 2 255

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_013230, AK125531, L33930, BC064619, AK026603, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          cell activation
          regulation of cytokine-mediated signaling pathway
          response to molecule of bacterial origin
          pre-B cell differentiation
          leukocyte migration involved in inflammatory response
          positive regulation of T cell mediated immune response to tumor cell
          positive regulation of inflammatory response to antigenic stimulus
          positive regulation of B cell apoptotic process
          immune response
          cell adhesion
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          leukocyte cell-cell adhesion
          cell surface receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          neuromuscular synaptic transmission
          negative regulation of neuroblast proliferation
          apoptotic mitochondrial changes
          positive regulation of neuron projection development
          single organismal cell-cell adhesion
          cell migration
          regulation of cell-cell adhesion
          positive regulation of cell-cell adhesion
          apoptotic nuclear changes
          negative regulation of B cell proliferation
          neuron projection development
          T cell costimulation
          B cell receptor transport into membrane raft
          chemokine receptor transport out of membrane raft
          negative regulation of transforming growth factor beta3 production
          positive regulation of integrin activation
          positive regulation of cell adhesion mediated by integrin
          regulation of cell-cell adhesion mediated by integrin
          positive regulation of cell-cell adhesion mediated by integrin
          negative regulation of erythrocyte clearance
          homotypic cell-cell adhesion
          negative regulation of erythrocyte aggregation
          positive regulation of T cell homeostatic proliferation
          positive regulation of activated T cell proliferation
          regulation of phosphorylation
          cholesterol homeostasis
          positive regulation of MAP kinase activity
          regulation of MAPK cascade
          response to estrogen
          regulation of B cell differentiation
          negative regulation of neuron differentiation
          respiratory burst
          negative regulation of T cell homeostatic proliferation
          positive regulation of alpha-beta T cell proliferation
          synaptic vesicle endocytosis
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of neurogenesis
          positive regulation of calcium-mediated signaling
          positive regulation of protein tyrosine kinase activity
          glomerular visceral epithelial cell differentiation
          glomerular parietal epithelial cell differentiation
          apoptotic signaling pathway
          positive regulation of nephron tubule epithelial cell differentiation
          protein kinase binding
          carbohydrate binding
          protein tyrosine kinase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline