Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UCN SIRNA Assays ›
Gene Symbol:
Ucn
Gene Name:
urocortin
Gene Aliases:
-
Chromosome Location:
Chr.6: 26602144 - 26602974 on Build Rnor_6.0
UniGene ID:
Rn.10190
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
46723
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ucn
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019150.1
NP_062023.1
2
398
GenBank mRNA
U33935.1
-
2
398
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_003353, BC104470, BC104471, CR542244.
This siRNA also targets the following transcripts in Mouse: NM_021290, BC144891, BC116948, BC116950.
Panther Classification:
Molecular Function -
peptide hormone
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
inflammatory response
response to oxidative stress
neuropeptide signaling pathway
female pregnancy
sensory perception of sound
learning or memory
feeding behavior
associative learning
aerobic respiration
positive regulation of gene expression
negative regulation of gene expression
pancreatic juice secretion
positive regulation of cell growth
positive regulation of cAMP biosynthetic process
negative regulation of histone deacetylation
neuron projection development
negative regulation of appetite
response to estradiol
positive regulation of interleukin-6 production
positive regulation of collagen biosynthetic process
positive regulation of peptidyl-serine phosphorylation
activation of protein kinase A activity
social behavior
gastric emptying
drinking behavior
negative regulation of apoptotic process
positive regulation of vascular permeability
positive regulation of translation
positive regulation of DNA replication
negative regulation of blood pressure
negative regulation of cell size
positive regulation of vasodilation
negative regulation of hormone secretion
response to pain
response to glucocorticoid
positive regulation of corticotropin secretion
regulation of synaptic transmission, glutamatergic
positive regulation of cardiac muscle contraction
negative regulation of gastric acid secretion
negative regulation of necrotic cell death
negative regulation of cell death
positive regulation of calcium ion import
negative regulation of neuron death
negative regulation of feeding behavior
positive regulation of behavioral fear response
G-protein coupled receptor binding
hormone activity
protein binding
histone deacetylase inhibitor activity
corticotropin-releasing hormone receptor 1 binding
corticotropin-releasing hormone receptor 2 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System