Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NKX2-5 SIRNA Assays ›
Gene Symbol:
Nkx2-5
Gene Name:
NK2 homeobox 5
Gene Aliases:
Csx, Nkx2.5
Chromosome Location:
Chr.10: 16635989 - 16638758 on Build Rnor_6.0
UniGene ID:
Rn.6179
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
46891
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nkx2-5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053651.1
NP_446103.1
1
341
GenBank mRNA
AF006664.1
-
1
341
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008700, L20300, X75415, BC139299, AK142146, and others.
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
vasculogenesis
heart looping
heart morphogenesis
outflow tract septum morphogenesis
outflow tract morphogenesis
cardiac conduction system development
atrioventricular node development
bundle of His development
Purkinje myocyte differentiation
cardiac ventricle morphogenesis
cardiac ventricle formation
right ventricular cardiac muscle tissue morphogenesis
ventricular trabecula myocardium morphogenesis
apoptotic process involved in heart morphogenesis
septum secundum development
proepicardium development
pulmonary myocardium development
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
heart development
adult heart development
positive regulation of cell proliferation
positive regulation of gene expression
negative regulation of cardiac muscle cell apoptotic process
positive regulation of transcription via serum response element binding
positive regulation of sodium ion transport
negative regulation of myotube differentiation
hemopoiesis
cell differentiation
BMP signaling pathway
thyroid gland development
response to estradiol
embryonic heart tube development
negative regulation of apoptotic process
sarcomere organization
positive regulation of neuron differentiation
positive regulation of heart contraction
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
spleen development
cardiac muscle tissue development
positive regulation of cardioblast differentiation
ventricular cardiac myofibril assembly
cardiac muscle cell differentiation
cardiac muscle tissue morphogenesis
regulation of cardiac muscle contraction
pharyngeal system development
cardiac muscle cell proliferation
regulation of cardiac muscle cell proliferation
heart contraction
cardiac muscle contraction
canonical Wnt signaling pathway
positive regulation of transcription initiation from RNA polymerase II promoter
heart trabecula formation
ventricular septum morphogenesis
atrial septum morphogenesis
atrioventricular node cell development
atrioventricular node cell fate commitment
embryonic heart tube left/right pattern formation
cellular response to growth factor stimulus
cardiovascular system development
negative regulation of canonical Wnt signaling pathway
positive regulation of voltage-gated calcium channel activity
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription cofactor activity
transcriptional activator activity, RNA polymerase II transcription factor binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
serum response element binding
protein homodimerization activity
histone deacetylase binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0