Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PFKM SIRNA Assays ›
Gene Symbol:
PFKM
Gene Name:
phosphofructokinase, muscle
Gene Aliases:
ATP-PFK, GSD7, PFK-1, PFK1, PFKA, PFKX, PPP1R122
Chromosome Location:
Chr.12: 48105278 - 48146404 on Build GRCh38
UniGene ID:
Hs.75160
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
46
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PFKM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000289.5
NP_000280.1
19, 20
2073
NM_001166686.1
NP_001160158.1
21, 22
2375
NM_001166687.1
NP_001160159.1
19, 20
1927
NM_001166688.1
NP_001160160.1
18, 19
2016
XM_005268974.1
XP_005269031.1
22, 23
2272
XM_005268975.1
XP_005269032.1
22, 23
2411
XM_005268976.3
XP_005269033.1
22, 23
2454
XM_005268977.1
XP_005269034.1
21, 22
2258
XM_005268978.2
XP_005269035.1
21, 22
2171
XM_005268979.1
XP_005269036.1
21, 22
2258
XM_011538487.1
XP_011536789.1
20, 21
2113
XM_011538488.2
XP_011536790.1
19, 20
2038
XM_017019467.1
XP_016874956.1
22, 23
2341
XM_017019468.1
XP_016874957.1
21, 22
2193
XM_017019469.1
XP_016874958.1
20, 21
2165
GenBank mRNA
AK126020.1
-
23, 24
2728
AK126229.1
-
22, 23
2325
AK303681.1
-
14, 15
1420
AK308185.1
-
14, 15
1396
AK312284.1
-
19, 20
1955
BC000534.2
-
18, 19
1903
2964710
BC007798.2
-
17, 18
1790
BC012799.2
-
18, 19
1903
2964710
BC013298.2
-
18, 19
1903
2964710
BC021203.2
-
19, 20
1925
4100117
CR541698.1
-
9, 10
943
J05533.1
-
10, 11
984
KU178251.1
-
18, 19
1888
KU178252.1
-
17, 18
1795
KU178253.1
-
17, 18
1723
KU178254.1
-
7, 8
733
M26066.1
-
19, 20
1964
U24183.1
-
18, 19
2216
Y00698.1
-
18, 19
1907
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00175997_m1
Panther Classification:
Molecular Function -
carbohydrate kinase
kinase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
glycogen catabolic process
fructose 6-phosphate metabolic process
glycolytic process
positive regulation of insulin secretion
glucose homeostasis
muscle cell cellular homeostasis
carbohydrate phosphorylation
protein oligomerization
canonical glycolysis
glycolysis from storage polysaccharide through glucose-1-phosphate
6-phosphofructokinase activity
protein binding
ATP binding
protein C-terminus binding
kinase binding
identical protein binding
protein homodimerization activity
metal ion binding
fructose binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-vvmmd:80/10.239.160.160:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0