Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CXCR4 SIRNA Assays ›
          Gene Symbol:
          CXCR4
          Gene Name:
          C-X-C motif chemokine receptor 4
          Gene Aliases:
          CD184, D2S201E, FB22, HM89, HSY3RR, LAP-3, LAP3, LCR1, LESTR, NPY3R, NPYR, NPYRL, NPYY3R, WHIM, WHIMS
          Chromosome Location:
          Chr.2: 136114349 - 136118155 on Build GRCh38
          UniGene ID:
          Hs.593413
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 4757

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CXCR4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008540.1 NP_001008540.1 1 399
          NM_003467.2 NP_003458.1 2 178
          GenBank mRNA AF025375.1 - 1 83
          AF147204.1 - 1 437
          AF348491.1 - 2 126
          AK129916.1 - 2 172
          AK296674.1 - 2 172
          AK309641.1 - 1 327
          AK309885.1 - 2 277
          AK312244.1 - 2 172
          AY242129.1 - 1 83
          BC020968.2 - 2 136 3846345
          BT006660.1 - 1 83
          D10924.1 - 2 159
          KU245646.1 - 2 296
          KU245647.1 - 2 182
          L01639.1 - 2 108
          L06797.1 - 2 155
          M99293.1 - 2 167
          X71635.1 - 2 150

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          response to hypoxia
          neuron migration
          epithelial cell development
          dendritic cell chemotaxis
          apoptotic process
          inflammatory response
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          neuron recognition
          response to virus
          fusion of virus membrane with host plasma membrane
          calcium-mediated signaling
          telencephalon cell migration
          entry into host cell
          regulation of cell migration
          regulation of programmed cell death
          myelin maintenance
          positive regulation of oligodendrocyte differentiation
          regulation of chemotaxis
          regulation of calcium ion transport
          cardiac muscle contraction
          chemokine-mediated signaling pathway
          cellular response to cytokine stimulus
          virus receptor activity
          actin binding
          G-protein coupled receptor activity
          protein binding
          drug binding
          coreceptor activity
          C-X-C chemokine receptor activity
          cytokine binding
          ubiquitin protein ligase binding
          myosin light chain binding
          ubiquitin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0