Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ctrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012536.1 | NP_036668.1 | 2, 3 | 164 | ||
GenBank mRNA | FQ232114.1 | - | 2, 3 | 186 | ||
FQ233595.1 | - | 2, 3 | 186 | |||
FQ234337.1 | - | 2, 3 | 186 | |||
FQ234339.1 | - | 2, 3 | 186 |